Order Entry
Slovenia
ContactUsLinkComponent
Anti-OTUB Mouse Monoclonal Antibody [clone: 1C12]
Anti-OTUB Mouse Monoclonal Antibody [clone: 1C12]
  ABNOH00055611-M09
 :  Abnova
undefined
Anti-OTUB Mouse Monoclonal Antibody [clone: 1C12]
  ABNOH00055611-M09
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    OTU Domain ubiquitin aldehyde binding 1
  • Antigen symbol:
    OTUB
  • Clonality:
    Monoclonal
  • Clone:
    1C12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    55611
  • Antigen synonyms:
    otubain 1|OTUB1|MGC111158|ubiquitin aldehyde binding 1|ubiquitin-specific protease otubain 1|HSPC263|OTU1|OTB1|MGC4584|FLJ20113|OTU-domain Ubal-binding 1|FLJ40710|OTU domain
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: OTUB
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human