Anti-PLA2G16 Mouse Monoclonal Antibody [clone: 4C4-2A11]
ABNOH00011145-M02
: Abnova
- Antibody type:Primary
- Antigen name:Phospholipase A2 Group XVI
- Antigen symbol:PLA2G16
- Clonality:Monoclonal
- Clone:4C4-2A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11145
- Antigen synonyms:HREV107-3|HRSL3|HREV107|HRASLS3|AdPLA|HREV107-1|H-REV107-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PLA2G16
Clonality: Monoclonal
Clone: 4C4-2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human