Anti-UBE4B Mouse Monoclonal Antibody [clone: 8F9]
Catalog # ABNOH00010277-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitination Factor E4B
- Antigen symbol:UBE4B
- Clonality:Monoclonal
- Clone:8F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10277
- Antigen synonyms:Ubiquitin-fusion degradation protein 2|KIAA0684|Homozygously deleted in neuroblastoma-1|HDNB1|Ubiquitin conjugation factor E4 B|UFD2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UBE4B
Clonality: Monoclonal
Clone: 8F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human