Anti-FZD3 Mouse Monoclonal Antibody [clone: 1B8]
ABNOH00007976-M02
: Abnova
- Antibody type:Primary
- Antigen name:Frizzled 3
- Antigen symbol:FZD3
- Clonality:Monoclonal
- Clone:1B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7976
- Antigen synonyms:Fz-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: FZD3
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human