Anti-UCK2 Mouse Monoclonal Antibody [clone: 1G12]
Catalog # ABNOH00007371-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Uridine-cytidine kinase 2
- Antigen symbol:UCK2
- Clonality:Monoclonal
- Clone:1G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:7371
- Antigen synonyms:Cytidine monophosphokinase 2|UMPK|TSA903|Uridine monophosphokinase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UCK2
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human