Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-UMOD Mouse Polyclonal Antibody
  ABNOH00007369-B01P
 :  Abnova
undefined
Anti-UMOD Mouse Polyclonal Antibody
  ABNOH00007369-B01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Uromodulin
  • Antigen symbol:
    UMOD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    7369
  • Antigen synonyms:
    MCKD2|FJHN|THGP|THP|ADMCKD2|HNFJ1|HNFJ
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

 

 

Type: Primary
Antigen: UMOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human