Anti-KLF10 Mouse Monoclonal Antibody [clone: 3E11]
ABNOH00007071-M12
: Abnova
- Antibody type:Primary
- Antigen name:Kruppel-like Factor 10
- Antigen symbol:KLF10
- Clonality:Monoclonal
- Clone:3E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7071
- Antigen synonyms:TIEG|TIEG1|EGR-alpha|EGRA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: KLF10
Clonality: Monoclonal
Clone: 3E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human