Anti-TEAD4 Mouse Monoclonal Antibody [clone: 1D10]
Catalog # ABNOH00007004-M12
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TEA domain family member 4
- Antigen symbol:TEAD4
- Clonality:Monoclonal
- Clone:1D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7004
- Antigen synonyms:TCF13L1|TEFR-1|EFTR-2|TEF-3|RTEF1|TEF3|hRTEF-1B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TEAD4
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human