Anti-PYCR1 Rabbit Polyclonal Antibody
Catalog # ABNOH00005831-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pyrroline-5-carboxylate reductase 1
- Antigen symbol:PYCR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5831
- Antigen synonyms:PIG45|ARCL3B|P5CR|PYCR|P5C|PP222|PRO3|ARCL2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PYCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human