Anti-MEF2BNB-MEF2B Mouse Monoclonal Antibody [clone: 4B5]
Catalog # ABNOH00004207-M24A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:MEF2BNB-MEF2B readthrough
- Antigen symbol:MEF2BNB-MEF2B
- Clonality:Monoclonal
- Clone:4B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4207
- Antigen synonyms:LOC729991-MEF2B|MEF2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: MEF2BNB-MEF2B
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human