Order Entry
Slovenia
ContactUsLinkComponent
Anti-CFHR1 Mouse Monoclonal Antibody [clone: 4D7]
Anti-CFHR1 Mouse Monoclonal Antibody [clone: 4D7]
  ABNOH00003078-M01
 :  Abnova
undefined
Anti-CFHR1 Mouse Monoclonal Antibody [clone: 4D7]
  ABNOH00003078-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Complement factor H-related 1
  • Antigen symbol:
    CFHR1
  • Clonality:
    Monoclonal
  • Clone:
    4D7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    3078
  • Antigen synonyms:
    H36-1|CFHL1P|HFL2|H36-2|CFHL1|HFL1|CFHL|CFHR1P|FHR1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: CFHR1
Clonality: Monoclonal
Clone: 4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human