Anti-MSR1 Mouse Monoclonal Antibody [clone: OTI9E5]
ORIGTA802777
: OriGene
- Antibody type:Primary
- Antigen name:macrophage scavenger receptor 1
- Antigen symbol:MSR1
- Clonality:Monoclonal
- Clone:OTI9E5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Amino acid number:197 to 451
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:42.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: MSR1 (macrophage scavenger receptor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human