Anti-APP Mouse Monoclonal Antibody [clone: OTI7B8]
Catalog # ORIGTA700259
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Amyloid Beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Monoclonal
- Clone:OTI7B8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Antigen synonyms:ABETA|beta Amyloid
- Storage buffer:Stored in PBS (pH 7,4) with 0,05% sodium azide, 10mg/ml BSA, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Recommended Dilutions: 1:250-1:1000
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human