Anti-SMS Mouse Monoclonal Antibody [clone: OTI1G7]
ORIGTA700119
: OriGene
- Antibody type:Primary
- Antigen name:spermine synthase
- Clonality:Monoclonal
- Clone:OTI1G7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) with 0,05% sodium azide, 10mg/ml BSA, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Recommended Dilutions: 1:250-1:1000
Type: Primary
Antigen: SMS (spermine synthase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human