Anti-LCMT1 Mouse Monoclonal Antibody [clone: OTI4A5]
Catalog # ORIGTA700110
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:leucine carboxyl methyltransferase 1
- Antigen symbol:LCMT1
- Clonality:Monoclonal
- Clone:OTI4A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) with 0,05% sodium azide, 10mg/ml BSA, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
- Recommended Dilutions: 1:250-1:1000
Type: Primary
Antigen: LCMT1 (leucine carboxyl methyltransferase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human