Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C11]
Catalog # ORIGTA600252
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Cystatin C
- Antigen symbol:CST3
- Clonality:Monoclonal
- Clone:OTI2C11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) containing 0,05% sodium azide and up to 5% trehalose
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: 1:100-1:1000
Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human