Anti-PLK1 Mouse Monoclonal Antibody [clone: 8C12]
Catalog # ORIGTA600111
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Polo-like kinase 1
- Antigen symbol:PLK1
- Clonality:Monoclonal
- Clone:OTI8C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) containing 0,05% sodium azide and up to 5% trehalose
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: 1:100-1:1000
Type: Primary
Antigen: PLK1 (polo-like kinase 1)
Clonality: Monoclonal
Clone: 8C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human