Anti-LECT2 Mouse Monoclonal Antibody [clone: OTI2A11]
Catalog # ORIGTA504091
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:leukocyte cell-derived chemotaxin 2
- Antigen symbol:LECT2
- Clonality:Monoclonal
- Clone:OTI2A11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:19 to 151
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:14.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150
Type: Primary
Antigen: LECT2 (leukocyte cell-derived chemotaxin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human