Anti-FABP2 Mouse Monoclonal Antibody [clone: OTI1B2]
ORIGTA503549
: OriGene
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 2, Intestinal
- Antigen symbol:FABP2
- Clonality:Monoclonal
- Clone:OTI1B2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:15.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: FABP2 (fatty acid binding protein 2, intestinal)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human