Anti-LCMT1 Mouse Monoclonal Antibody [clone: OTI2C9]
ORIGTA503125
: OriGene
- Antibody type:Primary
- Antigen name:leucine carboxyl methyltransferase 1
- Antigen symbol:LCMT1
- Clonality:Monoclonal
- Clone:OTI2C9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:38.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
- Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150
Type: Primary
Antigen: LCMT1 (leucine carboxyl methyltransferase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: