Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-TBC1D4 Mouse Monoclonal Antibody [clone: OTI5A6]
Catalog # ORIGTA502707
Supplier:  OriGene
undefined
Anti-TBC1D4 Mouse Monoclonal Antibody [clone: OTI5A6]
Catalog # ORIGTA502707
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TBC1 domain family, member 4
  • Antigen symbol:
    TBC1D4
  • Clonality:
    Monoclonal
  • Clone:
    OTI5A6
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    146.4 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TBC1D4 (TBC1 domain family, member 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human