Order Entry
Slovenia
ContactUsLinkComponent
Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI2F9]
Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI2F9]
Catalog # ORIGTA502586
Supplier:  OriGene
undefined
Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI2F9]
Catalog # ORIGTA502586
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lactamase, beta 2
  • Antigen symbol:
    LACTB2
  • Clonality:
    Monoclonal
  • Clone:
    OTI2F9
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    78 to 288
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    32.6 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LACTB2 (lactamase, beta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: