Anti-SDR9C7 Mouse Monoclonal Antibody [clone: OTI7F9]
Catalog # ORIGTA501733
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Short Chain Dehydrogenase/Reductase Family 9C Member 7
- Antigen symbol:SDR9C7
- Clonality:Monoclonal
- Clone:OTI7F9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SDR9C7 (short chain dehydrogenase/reductase family 9C, member 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human