Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI5D4]
Catalog # ORIGTA500936
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Fumarylacetoacetate hydrolase domain containing 2A
- Antigen symbol:FAHD2A
- Clonality:Monoclonal
- Clone:OTI5D4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:34.6 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: FAHD2A (fumarylacetoacetate hydrolase domain containing 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human