Anti-PDX1 Mouse Monoclonal Antibody [clone: OTI2A12]
Catalog # ORIGTA500038
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Pancreatic and duodenal homeobox 1
- Antigen symbol:PDX1
- Clonality:Monoclonal
- Clone:OTI2A12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:30.6 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:20000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100
Type: Primary
Antigen: PDX1 (pancreatic and duodenal homeobox 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human