Anti-MRGPRX2 Rabbit Polyclonal Antibody
Catalog # ORIGTA317037
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:MAS-related GPR, member X2
- Antigen symbol:MRGPRX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
Specifications
About this item
Recommended Dilutions: Immunochemistry: 20 µg/ml
Type: Primary
Antigen: MRGPRX2 (MAS-related GPR, member X2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human