Anti-COL1A1 Rabbit Antibody
Catalog # ORIGTA309096
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:collagen, type I, alpha 1
- Antigen symbol:COL1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Storage buffer:100 ul in PBS
- Molecular weight:180 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: COL1A1 (collagen, type I, alpha 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse