Anti-SLFN12 Rabbit Polyclonal Antibody
ORIGTA306894
: OriGene
- Antibody type:Primary
- Antigen name:schlafen family member 12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography via peptide column
- Pk:100 µG
- Recommended Dilutions: Western Blot: 1-2 μg/ml
Type: Primary
Antigen: SLFN12 (schlafen family member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human