Anti-RGP1 Rabbit Polyclonal Antibody
Catalog # ORIGTA306563
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:RGP1 retrograde golgi transport homolog (S. cerevisiae)
- Antigen symbol:RGP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography via peptide column
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: RGP1 (RGP1 retrograde golgi transport homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat