Order Entry
Slovenia
ContactUsLinkComponent
Anti-TMC6 Rabbit Polyclonal Antibody
Anti-TMC6 Rabbit Polyclonal Antibody
  ORIGTA306554
 :  OriGene
Anti-TMC6 Rabbit Polyclonal Antibody
  ORIGTA306554
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    transmembrane channel-like 6
  • Antigen symbol:
    TMC6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS containing 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography via peptide column
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: TMC6 (transmembrane channel-like 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat