Order Entry
Slovenia
ContactUsLinkComponent
Anti-NANOG Goat Polyclonal Antibody
Anti-NANOG Goat Polyclonal Antibody
Catalog # ORIGTA303048
Supplier:  OriGene
Anti-NANOG Goat Polyclonal Antibody
Catalog # ORIGTA303048
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nanog homeobox
  • Antigen symbol:
    NANOG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
  • Sequence:
    C-QNQRMKSKRWQKNN
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide. Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml Immunofluorescence: 5?g/ml

Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human