Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP615Y]
Catalog # ORIGTA300580
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 4
- Antigen symbol:MAP2K4
- Clonality:Monoclonal
- Clone:EP615Y
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS 49%,Sodium azide 0,01%,Glycerol 50%,BSA 0,05%
- Molecular weight:44 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:1000
Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat