Anti-KRT8 Rabbit Monoclonal Antibody [clone: E431-2]
Catalog # ORIGTA300410
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:keratin 8
- Antigen symbol:KRT8
- Clonality:Monoclonal
- Clone:E431-2
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS 49%,Sodium azide 0,01%,Glycerol 50%,BSA 0,05%
- Molecular weight:55 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:10000; Immunochemistry/Immunofluorescence: 1:250
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse