Anti-tdTomato Mouse Monoclonal Antibody
Catalog # ORIGTA180009
Supplier: OriGene
Specifications
- Antibody type:Tag
- Antigen name:tdTomato
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Tag
Antigen: tdTomato
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: