Anti-PKN beta Rabbit Polyclonal Antibody
ABGEAP7935A
: Abgent
- Antibody type:Primary
- Antigen name:Protein Kinase beta
- Antigen symbol:PKNB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (477–492 aa)
- Cross adsorption:No
- Gene ID:274
- Antigen synonyms:box dependant MYC interacting protein 1|myc box-dependent-interacting protein 1|box-dependent myc-interacting protein 1|amphiphysin II|AMPH2|AMPHL|SH3P9|amphiphysin-like protein
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:PSQFEAPGPFSEQASC
- Storage temperature:Cool
- Concentration:0,45 mg/ml
- Shipping temperature:Ice
- Immunogen:This BIN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 360-389 amino acids from the Central region of human BIN1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: PKN beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human