Order Entry
Slovenia
ContactUsLinkComponent
Anti-SLC47A1 Rabbit Polyclonal Antibody
Anti-SLC47A1 Rabbit Polyclonal Antibody
  ABGEAP5354B
 :  Abgent
undefined
Anti-SLC47A1 Rabbit Polyclonal Antibody
  ABGEAP5354B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 47 Member A1
  • Antigen symbol:
    SLC47A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SLC47A1 (solute carrier family 47 (multidrug and toxin extrusion), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human