Anti-LC3 (APG8C) Rabbit Polyclonal Antibody
Catalog # ABGEAP1804E
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Microtubule-associated proteins 1A/1B light chain 3C
- Antigen symbol:MAP1LC3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:MAP1LC3C|Autoph|MAP1A/MAP1B LC3 C|Microtubule-associated proteins 1A/1B light chain 3C|MAP1A/1B light chain 3 C|MAP1 light chain 3-like protein 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: LC3 (APG8C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human