Order Entry
Slovenia
ContactUsLinkComponent
Anti-SIGLEC7 (D-siglec) Rabbit Polyclonal Antibody
Anti-SIGLEC7 (D-siglec) Rabbit Polyclonal Antibody
Catalog # ABGEAP1625A
Supplier:  Abgent
undefined
Anti-SIGLEC7 (D-siglec) Rabbit Polyclonal Antibody
Catalog # ABGEAP1625A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sialic acid binding Ig-like lectin 7
  • Antigen symbol:
    SIGLEC7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal ( 1–50 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    CD328|SIGLECP2|p75/AIRM1|p75|CDw328|D-siglec|SIGLEC19P|AIRM1|QA79|SIGLEC-7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SIGLEC7 (D-siglec)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse