Order Entry
Slovenia
ContactUsLinkComponent
Anti-MAP2K4 Rabbit Polyclonal Antibody
Anti-MAP2K4 Rabbit Polyclonal Antibody
  ABGEAP14920B
 :  Abgent
Anti-MAP2K4 Rabbit Polyclonal Antibody
  ABGEAP14920B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase Kinase 4
  • Antigen symbol:
    MAP2K4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Zebrafish,
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Gene ID:
    26398
  • Antigen synonyms:
    MKK4|SAPKK-1|PRKMK4|SAPKK1|MAPKK4|SERK1|SKK1|MEK4|JNKK1|JNKK|SEK1
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    44.114 kDa
  • Sequence:
    DPSASRQGYDVRSDC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,48 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Mouse Map2k4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 259-287 amino acids from the C-terminal region of mouse Map2k4.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse