Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-MAP3K6 Rabbit Polyclonal Antibody
  ABGEAP13918A
 :  Abgent
undefined
Anti-MAP3K6 Rabbit Polyclonal Antibody
  ABGEAP13918A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase Kinase Kinase 6
  • Antigen symbol:
    MAP3K6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    ASK2, MAPKKK6, MEKK6
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: MAP3K6 (mitogen-activated protein kinase kinase kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse