Anti-IRF4 Rabbit Polyclonal Antibody
Catalog # ABGEAP12637C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:IRF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: IRF4 (interferon regulatory factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human