Anti-SLC13A4 Rabbit Polyclonal Antibody
AVIVARP66472_P050
- Antibody type:Primary
- Antigen name:solute carrier family 13 (sodium/sulfate symporter), member 4
- Antigen symbol:Solute carrier family 13, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:26266
- Antigen synonyms:SUT-1|SUT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SLC13A4 (solute carrier family 13 (sodium/sulfate symporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human