Anti-LYAR Rabbit Polyclonal Antibody
AVIVARP57042_P050
- Antibody type:Primary
- Antigen name:Ly1 antibody reactive
- Antigen symbol:LYAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:55646
- Antigen synonyms:ZLYAR|ZC2HC2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LYAR (Ly1 antibody reactive)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: