Anti-GP2 Rabbit Polyclonal Antibody
Catalog # AVIVARP44543_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein 2
- Antigen symbol:GP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:2813
- Antigen synonyms:ZAP75
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GP2 (glycoprotein 2 (zymogen granule membrane))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: