Anti-SLC7A1 Rabbit Polyclonal Antibody
Catalog # AVIVARP43837_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
- Antigen symbol:Solute carrier family 7, member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6541
- Antigen synonyms:ATRC1|REC1L|HCAT1|CAT-1|ERR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC7A1 (solute carrier family 7 (cationic amino acid transporter, y+ system), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: