Anti-SLC1A4 Rabbit Polyclonal Antibody
Catalog # AVIVARP43827_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 1 (glutamate/neutral amino acid transporter), member 4
- Antigen symbol:Solute carrier family 1, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:6509
- Antigen synonyms:ASCT1, SATT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC1A4 (solute carrier family 1 (glutamate/neutral amino acid transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: