Anti-SLC7A3 Rabbit Polyclonal Antibody
AVIVARP43806_P050
- Antibody type:Primary
- Antigen name:solute carrier family 7 (cationic amino acid transporter, y+ system), member 3
- Antigen symbol:Solute carrier family 7, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:11989
- Antigen synonyms:CAT-3|SLC7A1|CAT3|SLC7A2|Atrc3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SLC7A3 (solute carrier family 7 (cationic amino acid transporter, y+ system), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: