Anti-BMP2K Rabbit Polyclonal Antibody
Catalog # AVIVARP42483_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:BMP2 inducible kinase
- Antigen symbol:BMP2K
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55589
- Antigen synonyms:HRIHFB2017|BIKE
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: BMP2K (BMP2 inducible kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: