Anti-UPB1 Rabbit Polyclonal Antibody
AVIVARP42467_T100
- Antibody type:Primary
- Antigen name:Ureidopropionase, beta
- Antigen symbol:UPB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:51733
- Antigen synonyms:BUP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: UPB1 (ureidopropionase, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: