Anti-PSG1 Rabbit Polyclonal Antibody
AVIVARP42325_T100
- Antibody type:Primary
- Antigen name:pregnancy specific beta-1-glycoprotein 1
- Antigen symbol:PSG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5669
- Antigen synonyms:PSBG1|PSG95|PS-beta-C/D|DHFRP2|CD66f|PSBG-1|PSGIIA|SP1|PSGGA|B1G1|FL-NCA-1/2|PS-beta-G-1|PBG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: PSG1 (pregnancy specific beta-1-glycoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human